Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STYX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193128.1 | NP_001180057.1 | 8 - 9 | 697 | 119 | |
GenBank mRNA | BC107547.1 | - | 6 - 7 | 1193 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |