Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL26A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193154.1 | NP_001180083.1 | 9 - 10 | 1109 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |