Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193169.2 | NP_001180098.1 | 1 - 2 | 71 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |