Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZKSCAN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193173.1 | NP_001180102.1 | 3 - 4 | 1061 | 81 | |
GenBank mRNA | BC149240.1 | - | 4 - 5 | 1221 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |