Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193197.3 | NP_001180126.3 | 5 - 6 | 1313 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Probe spans exons |