Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193248.3 | NP_001180177.1 | 6 - 7 | 736 | 98 | |
GenBank mRNA | BC104517.1 | - | 6 - 7 | 628 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |