Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193259.1 | NP_001180188.1 | 2 - 3 | 234 | 85 | |
GenBank mRNA | BC149735.1 | - | 2 - 3 | 243 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |