Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMSTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195011.1 | NP_001181940.1 | 8 - 9 | 1010 | 75 | |
GenBank mRNA | BC151712.1 | - | 7 - 8 | 953 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |