Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195064.1 | NP_001181993.1 | 1 - 2 | 52 | 59 | |
GenBank mRNA | BC114898.1 | - | 1 - 2 | 52 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |