Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCA5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195068.1 | NP_001181997.1 | 4 - 5 | 1183 | 83 | |
GenBank mRNA | BC151815.1 | - | 3 - 4 | 1009 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |