Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GALNAC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001151.2 | NP_001001151.2 | 1 - 2 | 150 | 95 | |
GenBank mRNA | AJ620949.1 | - | 1 - 2 | 78 | 95 | |
BC123467.1 | - | 1 - 2 | 150 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |