Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARTPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007820.3 | NP_001007821.1 | 2 - 3 | 264 | 86 | |
GenBank mRNA | BC114792.1 | - | 2 - 3 | 266 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |