Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOLA-DOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013600.1 | NP_001013618.1 | 3 - 4 | 780 | 75 | |
GenBank mRNA | AB117945.1 | - | 3 - 4 | 641 | 75 | |
AB117946.1 | - | 3 - 4 | 780 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |