Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035447.2 | NP_001030524.1 | 2 - 3 | 327 | 92 | |
GenBank mRNA | BC103337.1 | - | 2 - 3 | 328 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |