Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038045.2 | NP_001033134.2 | 2 - 3 | 256 | 115 | |
GenBank mRNA | BC109746.1 | - | 2 - 3 | 251 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |