Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046158.2 | NP_001039623.2 | 13 - 14 | 1746 | 86 | |
GenBank mRNA | BC112851.1 | - | 12 - 13 | 1847 | 86 | |
BT025363.1 | - | 13 - 14 | 1674 | 86 | ||
EV648317.1 | - | 3 - 4 | 291 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |