Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GORASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046518.2 | NP_001039983.1 | 7 - 8 | 1009 | 90 | |
GenBank mRNA | BC110126.1 | - | 7 - 8 | 1010 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |