Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPV17L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046602.2 | NP_001040067.1 | 3 - 4 | 442 | 100 | |
GenBank mRNA | BC112608.1 | - | 3 - 4 | 444 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |