Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSSC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075410.1 | NP_001068878.1 | 2 - 3 | 185 | 58 | |
GenBank mRNA | BC116073.1 | - | 2 - 3 | 185 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |