Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076055.3 | NP_001069523.3 | 4 - 5 | 1263 | 63 | |
GenBank mRNA | DV880013.1 | - | 3 - 4 | 515 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Bos taurus Probe spans exons |