Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198996.1 | NP_001185925.1 | 1 - 2 | 287 | 87 | |
GenBank mRNA | AY911363.1 | - | 1 - 2 | 115 | 87 | |
EH164738.1 | - | 1 - 2 | 115 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Bos taurus Probe spans exons |