Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205307.3 | NP_001192236.2 | 25 - 26 | 7236 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |