Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205320.1 | NP_001192249.1 | 5 - 6 | 707 | 71 | |
GenBank mRNA | DV894107.1 | - | 2 - 3 | 146 | 71 | |
EV612341.1 | - | 5 - 6 | 707 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Bos taurus Probe spans exons |