Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205375.2 | NP_001192304.1 | 4 - 5 | 609 | 59 | |
GenBank mRNA | BC109767.1 | - | 4 - 5 | 610 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |