Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205380.1 | NP_001192309.1 | 1 - 2 | 187 | 102 | |
GenBank mRNA | EE235151.1 | - | 1 - 2 | 187 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |