Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205382.1 | NP_001192311.1 | 1 - 2 | 499 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |