Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099387.2 | NP_001092857.1 | 1 | 1422 | 83 | |
GenBank mRNA | BC146032.1 | - | 1 | 1423 | 83 | |
BC151574.1 | - | 1 | 1385 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |