Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270317.4 | NP_001257246.2 | 28 - 29 | 8099 | 98 | |
NM_001373583.1 | NP_001360775.1 | 28 - 29 | 8066 | 98 | ||
NM_001381151.1 | NP_001368758.1 | 28 - 29 | 8082 | 98 | ||
NM_001381152.1 | NP_001368759.1 | 28 - 29 | 8082 | 98 | ||
NM_001381153.1 | NP_001368756.1 | 28 - 29 | 8049 | 98 | ||
NM_001381154.1 | NP_001368757.1 | 28 - 29 | 8049 | 98 | ||
NM_001381155.1 | NP_001368146.1 | 28 - 29 | 8076 | 98 | ||
NM_001381156.2 | NP_001368760.1 | 28 - 29 | 8072 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |