Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csq-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078037.7 | NP_510438.1 | 3 - 4 | 338 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |