Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hch-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078039.7 | NP_510440.1 | 3 - 4 | 582 | 107 | |
GenBank mRNA | D85744.1 | - | 3 - 4 | 683 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |