Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078052.5 | NP_510453.1 | 9 - 10 | 1125 | 66 | |
GenBank mRNA | AF052060.1 | - | 9 - 10 | 1155 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |