Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078059.6 | NP_510460.2 | 4 - 5 | 636 | 91 | |
GenBank mRNA | AY064464.1 | - | 4 - 5 | 649 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |