Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078059.6 | NP_510460.2 | 8 - 9 | 1999 | 70 | |
GenBank mRNA | AY064464.1 | - | 8 - 9 | 2012 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |