Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dsc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078096.4 | NP_510497.1 | 3 - 4 | 592 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |