Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078107.6 | NP_510508.1 | 4 - 5 | 465 | 117 | |
GenBank mRNA | AB012700.1 | - | 4 - 5 | 487 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |