Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270326.3 | NP_001257255.1 | 4 - 5 | 395 | 77 | |
NM_001270327.3 | NP_001257256.1 | 2 - 3 | 114 | 77 | ||
NM_001350352.4 | NP_001337313.1 | 3 | 390 | 77 | ||
NM_001350353.4 | NP_001337312.1 | 3 | 390 | 77 | ||
NM_001350354.3 | NP_001337317.1 | 3 - 4 | 219 | 77 | ||
NM_001350355.3 | NP_001337316.1 | 3 - 4 | 141 | 77 | ||
NM_001350356.3 | NP_001337319.1 | 5 | 571 | 77 | ||
NM_001350357.3 | NP_001337318.1 | 3 - 4 | 180 | 77 | ||
NM_001350358.2 | NP_001337320.1 | 4 - 5 | 395 | 77 | ||
GenBank mRNA | Z19122.1 | - | 416 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |