Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001364052.5 | NP_001350995.1 | 9 | 2111 | 65 | |
NM_001364054.4 | NP_001350997.1 | 10 - 11 | 2126 | 65 | ||
NM_001368560.4 | NP_001355485.1 | 9 | 2153 | 65 | ||
NM_078145.5 | NP_510546.1 | 8 - 9 | 2947 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |