Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sprr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078154.6 | NP_510555.2 | 9 - 10 | 1487 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |