Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270335.2 | NP_001257264.1 | 6 - 7 | 1217 | 133 | |
NM_001270336.3 | NP_001257265.1 | 5 - 6 | 1014 | 133 | ||
NM_001313436.5 | NP_001300365.1 | 7 - 8 | 1164 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |