Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srg-64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078188.2 | NP_510589.2 | 4 - 5 | 831 | 165 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |