Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375198.2 | NP_001361896.1 | 5 - 6 | 748 | 89 | |
NM_001375199.1 | NP_001361830.1 | 5 - 6 | 774 | 89 | ||
NM_001375200.2 | NP_001361897.1 | 4 - 5 | 563 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |