Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375198.2 | NP_001361896.1 | 7 - 8 | 1152 | 60 | |
NM_001375199.1 | NP_001361830.1 | 7 - 8 | 1178 | 60 | ||
NM_001375200.2 | NP_001361897.1 | 6 - 7 | 967 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |