Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029217.6 | NP_001024388.1 | 10 - 11 | 1237 | 126 | |
NM_001029218.7 | NP_001024389.1 | 7 - 8 | 1291 | 126 | ||
GenBank mRNA | AY305831.1 | - | 8 - 9 | 1043 | 126 | |
AY305832.1 | - | 8 - 9 | 1033 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |