Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078280.8 | NP_510681.4 | 20 - 21 | 4435 | 67 | |
GenBank mRNA | DQ360811.1 | - | 20 - 21 | 3719 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |