Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078280.8 | NP_510681.4 | 4 - 5 | 1151 | 89 | |
GenBank mRNA | DQ360811.1 | - | 4 - 5 | 435 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |