Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029682.5 | NP_001024853.1 | 8 - 9 | 882 | 86 | |
NM_001029684.6 | NP_001024855.1 | 7 - 8 | 764 | 86 | ||
NM_001381184.1 | NP_001367802.1 | 6 - 7 | 672 | 86 | ||
GenBank mRNA | AY204163.1 | - | 7 - 8 | 748 | 86 | |
AY204164.1 | - | 7 - 8 | 762 | 86 | ||
AY204165.1 | - | 7 - 8 | 777 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |