Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lec-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078443.8 | NP_510844.1 | 2 - 3 | 340 | 81 | |
GenBank mRNA | AB038500.1 | - | 2 - 3 | 217 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |