Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306233.3 | NP_001293162.1 | 5 - 6 | 579 | 87 | |
NM_001380379.1 | NP_001367885.1 | 4 - 5 | 494 | 87 | ||
GenBank mRNA | AB096875.1 | - | 4 - 5 | 481 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |