Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wwp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306342.3 | NP_001293271.1 | 1 - 2 | 212 | 110 | |
NM_170794.7 | NP_740775.1 | 5 - 6 | 1089 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |