Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383770.1 | 6 - 7 | 950 | 64 | ||
NM_170806.5 | NP_740789.1 | 5 - 6 | 1027 | 64 | ||
GenBank mRNA | AY008139.1 | - | 5 - 6 | 722 | 64 | |
U56864.1 | - | 5 - 6 | 939 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |